ENA087844907 Enamine

5-methoxy-2-methyl-1-propyl-1H-indole-3-carboxylic acid

  • MDL number MFCD08246124
  • Cas Number : 17826-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171...173
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24260

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24260
Availability details for
Quantity
  • Availability
  • Add to Cart