ENA087844924 Enamine

1-[2-(1H-pyrazol-1-ylmethyl)phenyl]methanamine

  • MDL number MFCD08245275
  • Cas Number : 878466-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47...49
Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24281

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24281
Availability details for
Quantity
  • Availability
  • Add to Cart