ENA087844926 Enamine

1-(4-chlorobenzenesulfonyl)cyclopentane-1-carboxylic acid

  • MDL number MFCD03701697
  • Cas Number : 505071-95-4
  • Molecular Weight : 288.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-24283

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24283
Availability details for
Quantity
  • Availability
  • Add to Cart