ENA087844933 Enamine

3-(2,3-dihydro-1,4-benzodioxin-6-yl)propanoic acid

  • MDL number MFCD06823843
  • Cas Number : 14939-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-66
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24291

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24291
Availability details for
Quantity
  • Availability
  • Add to Cart