ENA087844935 Enamine

4-(2-mercapto-4-oxoquinazolin-3(4H)-yl)benzonitrile

  • MDL number MFCD08444232
  • Cas Number : 879361-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24298

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24298
Availability details for
Quantity
  • Availability
  • Add to Cart