ENA087844936 Enamine

2-[4-methyl-2-(methylsulfanyl)-1,3-thiazol-5-yl]acetic acid

  • MDL number MFCD08262837
  • Cas Number : 77446-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-24300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24300
Availability details for
Quantity
  • Availability
  • Add to Cart