ENA087844938 Enamine

3-[(phenylsulfanyl)methyl]furan-2-carboxylic acid

  • MDL number MFCD08245265
  • Cas Number : 878465-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24302

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24302
Availability details for
Quantity
  • Availability
  • Add to Cart