ENA087844941 Enamine

methyl 3-formyl-2-furoate

  • MDL number MFCD08245262
  • Cas Number : 28921-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24306

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24306
Availability details for
Quantity
  • Availability
  • Add to Cart