ENA087844943 Enamine

5-[(3aS,4S,6aR)-2,5,5-trioxo-hexahydro-1H-5lambda6-thieno[3,4-d]imidazol-4-yl]pentanoic acid

  • MDL number MFCD08444234
  • Cas Number : 40720-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24308
Availability details for
Quantity
  • Availability
  • Add to Cart