ENA087844949 Enamine

2-chloro-N-(4-chlorophenyl)-N-(2,2,2-trifluoroethyl)acetamide

  • MDL number MFCD08245279
  • Cas Number : 878466-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-24318

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24318
Availability details for
Quantity
  • Availability
  • Add to Cart