ENA087844950 Enamine

2-(8-chloro-2,3-dihydro-1,4-benzodioxin-6-yl)acetic acid

  • MDL number MFCD08245281
  • Cas Number : 878466-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24319

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24319
Availability details for
Quantity
  • Availability
  • Add to Cart