ENA087844958 Enamine

1-methyl-4-(piperazine-1-sulfonyl)-1H-pyrrole-2-carboxamide

  • MDL number MFCD08262839
  • Cas Number : 879362-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-24331

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24331
Availability details for
Quantity
  • Availability
  • Add to Cart