ENA087844961 Enamine

2,2,2-trifluoroethyl carbamate

  • MDL number MFCD00460713
  • Cas Number : 461-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42-43
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-24334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24334
Availability details for
Quantity
  • Availability
  • Add to Cart