ENA087844967 Enamine

1-[(5-carbamoyl-1-methyl-1H-pyrrol-3-yl)sulfonyl]piperidine-4-carboxylic acid

  • MDL number MFCD08262847
  • Cas Number : 879362-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24351

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24351
Availability details for
Quantity
  • Availability
  • Add to Cart