ENA087844969 Enamine

{[(3,4-diethoxyphenyl)sulfonyl]amino}acetic acid

  • MDL number MFCD08262846
  • Cas Number : 879362-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24354

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24354
Availability details for
Quantity
  • Availability
  • Add to Cart