ENA087844973 Enamine

methyl 1-cyclopropyl-5-hydroxy-2-methyl-1H-indole-3-carboxylate

  • MDL number MFCD08262836
  • Cas Number : 879362-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24361

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24361
Availability details for
Quantity
  • Availability
  • Add to Cart