ENA087844982 Enamine

2-chloro-N-[(4-methoxyphenyl)methyl]-N-[1-(thiophen-2-yl)propan-2-yl]acetamide

  • MDL number MFCD08261041
  • Cas Number : 879319-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-22905

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22905
Availability details for
Quantity
  • Availability
  • Add to Cart