ENA087844994 Enamine

5-methoxy-4-(2-methoxyethoxy)-2-nitrobenzoic acid

  • MDL number MFCD08262814
  • Cas Number : 879361-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24409

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24409
Availability details for
Quantity
  • Availability
  • Add to Cart