ENA087844995 Enamine

4-(5-formylfuran-2-yl)-N,N-dimethylbenzene-1-sulfonamide

  • MDL number MFCD06250084
  • Cas Number : 710294-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24410

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24410
Availability details for
Quantity
  • Availability
  • Add to Cart