ENA087844996 Enamine

2-{2-[(5-amino-1,3,4-thiadiazol-2-yl)thio]ethyl}-1H-isoindole-1,3(2H)-dione

  • MDL number MFCD04619679
  • Cas Number : 556024-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15462

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15462
Availability details for
Quantity
  • Availability
  • Add to Cart