ENA087845010 Enamine

N-phenyl-2-(phenylamino)acetamide

  • MDL number MFCD00965324
  • Cas Number : 2567-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24440

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24440
Availability details for
Quantity
  • Availability
  • Add to Cart