ENA087845015 Enamine

4-(2-chloropropanoyl)-3,4-dihydroquinoxalin-2(1H)-one

  • MDL number MFCD08245260
  • Cas Number : 878465-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23091

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23091
Availability details for
Quantity
  • Availability
  • Add to Cart