ENA087845019 Enamine

4-(4-fluorophenyl)-5-(furan-2-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD07774562
  • Cas Number : 259154-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-24454

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24454
Availability details for
Quantity
  • Availability
  • Add to Cart