ENA087845023 Enamine

3-imino-4-(4-methoxyphenyl)-1lambda6,2,5-thiadiazolidine-1,1-dione

  • MDL number MFCD02637033
  • Cas Number : 141542-12-3
  • Molecular Weight : 241.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24460

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24460
Availability details for
Quantity
  • Availability
  • Add to Cart