ENA087845035 Enamine

N-{4-[benzyl(methyl)sulfamoyl]phenyl}-2-chloroacetamide

  • MDL number MFCD08262820
  • Cas Number : 879361-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-23161

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23161
Availability details for
Quantity
  • Availability
  • Add to Cart