ENA087845036 Enamine

3-(fluorosulfonyl)benzoic acid

  • MDL number MFCD00007415
  • Cas Number : 454-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-155
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24489

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-24489
pictograms GHS05,GHS07
hstatements H302,H314,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart