ENA087845037 Enamine

2-chloro-N-[4-methyl-5-phenyl-3-(piperidine-1-carbonyl)thiophen-2-yl]acetamide

  • MDL number MFCD08261059
  • Cas Number : 879319-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23169

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23169
Availability details for
Quantity
  • Availability
  • Add to Cart