ENA087845038 Enamine

2-chloro-N-[2-(3-chlorophenyl)ethyl]acetamide

  • MDL number MFCD01733628
  • Cas Number : 34162-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23172
Availability details for
Quantity
  • Availability
  • Add to Cart