ENA087845042 Enamine

2-chloro-N-[1-(1-naphthyl)ethyl]acetamide

  • MDL number MFCD08239614
  • Cas Number : 131149-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 303...305
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23213

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23213
Availability details for
Quantity
  • Availability
  • Add to Cart