ENA087845043 Enamine

2-methyl-5-{[(2-methylphenyl)amino]sulfonyl}benzoic acid

  • MDL number MFCD03140662
  • Cas Number : 104941-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24503

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24503
Availability details for
Quantity
  • Availability
  • Add to Cart