ENA087845046 Enamine

2-chloro-N-[1-(2,3-dihydro-1,4-benzodioxin-6-yl)-2-methylpropyl]acetamide

  • MDL number MFCD08262842
  • Cas Number : 879362-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-23215

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23215
Availability details for
Quantity
  • Availability
  • Add to Cart