ENA087845048 Enamine

N-benzyl-2-(2-chloroacetamido)benzamide

  • MDL number MFCD01907034
  • Cas Number : 53824-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-18235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-18235
Availability details for
Quantity
  • Availability
  • Add to Cart