ENA087845052 Enamine

2-chloro-1-(chloromethyl)-4-fluorobenzene

  • MDL number MFCD00236026
  • Cas Number : 93286-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18254

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-18254
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart