ENA087845072 Enamine

N-(piperidin-4-yl)benzamide

  • MDL number MFCD01314226
  • Cas Number : 33953-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24697

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24697
Availability details for
Quantity
  • Availability
  • Add to Cart