ENA087845087 Enamine

2-chloro-N-[2-(phenylthio)phenyl]propanamide

  • MDL number MFCD08261040
  • Cas Number : 879319-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23359

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23359
Availability details for
Quantity
  • Availability
  • Add to Cart