ENA090493021 Enamine

1-(chloroacetyl)-4-(2-chlorophenyl)piperazine

  • MDL number MFCD01252305
  • Cas Number : 330601-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 245...247
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-22967

Safety Information

msds https://www.enaminestore.com/catalog/EN300-22967
Availability details for
Quantity
  • Availability
  • Add to Cart