ENA090493026 Enamine

2-chloro-1-[4-(4-ethoxybenzenesulfonyl)piperazin-1-yl]ethan-1-one

  • MDL number MFCD08444129
  • Cas Number : 923743-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23040

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23040
Availability details for
Quantity
  • Availability
  • Add to Cart