ENA090493029 Enamine

2-chloro-N-[3-(diethylsulfamoyl)-4-ethoxyphenyl]acetamide

  • MDL number MFCD08444142
  • Cas Number : 923773-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-23199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23199
Availability details for
Quantity
  • Availability
  • Add to Cart