ENA090493036 Enamine

4-[(5-methyl-1,2,4-oxadiazol-3-yl)methoxy]benzoic acid

  • MDL number MFCD08444265
  • Cas Number : 923696-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24467

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24467
Availability details for
Quantity
  • Availability
  • Add to Cart