ENA090493038 Enamine

4-cyclopropyl-3-(methylsulfonyl)-5-thien-2-yl-4H-1,2,4-triazole

  • MDL number MFCD08444276
  • Cas Number : 923767-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24496

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24496
Availability details for
Quantity
  • Availability
  • Add to Cart