ENA090493039 Enamine

2-[(5-methyl-1,2,4-oxadiazol-3-yl)methoxy]benzoic acid

  • MDL number MFCD08444277
  • Cas Number : 923204-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24497

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24497
Availability details for
Quantity
  • Availability
  • Add to Cart