ENA090493043 Enamine

3-(2-chloroacetamido)-N-cyclopropylbenzamide

  • MDL number MFCD08444299
  • Cas Number : 923678-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-24602

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24602
Availability details for
Quantity
  • Availability
  • Add to Cart