ENA090493047 Enamine

3-cyclopropyl-1-phenyl-1H-1,2,4-triazole-5-thiol

  • MDL number MFCD08444307
  • Cas Number : 923193-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-24689

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24689
Availability details for
Quantity
  • Availability
  • Add to Cart