ENA090493050 Enamine

2-imino-2,3-dihydro-1,3-thiazole-5-carboxamide

  • MDL number MFCD23146107
  • Cas Number : 52499-04-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-24707

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24707
Availability details for
Quantity
  • Availability
  • Add to Cart