ENA090493066 Enamine

2-[(morpholin-4-yl)methyl]quinazoline-4-thiol

  • MDL number MFCD08444331
  • Cas Number : 923711-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-25319

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25319
Availability details for
Quantity
  • Availability
  • Add to Cart