ENA090493068 Enamine

2-[(dimethylamino)methyl]quinazoline-4-thiol

  • MDL number MFCD08444334
  • Cas Number : 923140-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131...133
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25329

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25329
Availability details for
Quantity
  • Availability
  • Add to Cart