ENA090493081 Enamine

3-(5-formyl-2-furyl)-1-phenyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD08444357
  • Cas Number : 956769-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146...148
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25407

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25407
Availability details for
Quantity
  • Availability
  • Add to Cart