ENA090493083 Enamine

2-(1-methylidene-3-oxo-2,3-dihydro-1H-isoindol-2-yl)acetic acid

  • MDL number MFCD08444360
  • Cas Number : 62100-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-25415

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25415
Availability details for
Quantity
  • Availability
  • Add to Cart