ENA090493093 Enamine

4-(hydroxyimino)piperidine-1-carboxamide

  • MDL number MFCD08444373
  • Cas Number : 923106-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25437

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25437
Availability details for
Quantity
  • Availability
  • Add to Cart