ENA090493095 Enamine

N,N,1-trimethyl-3-oxo-1,2,3,4-tetrahydroquinoxaline-6-sulfonamide

  • MDL number MFCD08444375
  • Cas Number : 923128-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25439

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25439
Availability details for
Quantity
  • Availability
  • Add to Cart